Learn More
Apexbio Technology LLC ABT-199, 50mg. Cas: 1257044-40-8 MFCD: MFCD23160052. Bcl-2 inhibitor,potent and selective
SDP
Supplier:  Apexbio Technology LLC A819450
Encompass_Preferred
ABT-199, developed through a structure-based reverse engineering process, is a novel and specific inhibitor of B-cell lymphoma/leukemia 2 (BCL-2) maintaining a sub-nanomolar affinity towards BCL-2 and over three orders of magnitude less affinity towards BCL-XL Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5073