Learn More
Abcam ABT-199 (Venetocla x ), Bcl-2 inhibitor., 5MG
Catalog No. 502587957
Encompass_Preferred
MW 868.4 Da, Purity >98%. Potent, selective Bcl-2 inhibitor (Ki <0.01 nM) that inhibits the growth of Bcl-2–dependent tumors while sparing platelets. Bcl-2–specific BH3 mimetic. Promising agent in the treatment of chronic lymphocytic leukaemic and estrogen receptor-positive breast cancer.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-7957 Supplier Abcam Supplier No. AB2172985MG
MW 868.4 Da, Purity >98%. Potent, selective Bcl-2 inhibitor (Ki <0.01 nM) that inhibits the growth of Bcl-2–dependent tumors while sparing platelets. Bcl-2–specific BH3 mimetic. Promising agent in the treatment of chronic lymphocytic leukaemic and estrogen receptor-positive breast cancer.
The product is subject to the following: Abcam Restricted Use Statement