Learn More
Chemie Tek ABT-263, 923564-51-6, MFCD12756219, 10 mg
SDP
Supplier:  Chemie Tek CTA26310MG
Encompass_Preferred
Formula: C47H55ClF3N5O6S3, MW: 974.6, Synonym: Navitoclax. ABT-263 (Navitoclax) is a potent, selective and orally bioavailable inhibitor of B-cell lymphoma-2 (BCL-2) family proteins, binding potently to both BCL-2 and BCL-XL with Ki values of <1 nM.
Catalog No. NC0934746