Learn More
Selleck Chemical LLC Navitoclax (ABT-263) 25mg 923564-51-6
SDP
Catalog No. 508498
Encompass_Preferred
Navitoclax (ABT-263) is a potent inhibitor of Bcl-xL, Bcl-2 and Bcl-w with Ki of ? 0.5 nM, ?1 nM and ?1 nM in cell-free assays, but binds more weakly to Mcl-1 and A1. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-849-8 Supplier Selleck Chemical LLC Supplier No. S100125MG
Navitoclax (ABT-263) is a potent inhibitor of Bcl-xL, Bcl-2 and Bcl-w with Ki of ? 0.5 nM, ?1 nM and ?1 nM in cell-free assays, but binds more weakly to Mcl-1 and A1. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.