Learn More
Medchemexpress LLC ABT-325 1mg | 1mg
SDP
Catalog No. 5000325771
Encompass_Preferred
ABT-325 is a humanized antibody expressed in CHO that targets IL-18 ABT-325 has a huIgG1 type heavy chain and a hu type light chain with a predicted molecular weight (MW) of 146 28 kDa The isotype control for ABT-325 can refer to Human IgG1 kappa Isotype Control (HY-P99001)
Catalog No. 50-003-25771 Supplier Medchemexpress LLC Supplier No. HYP9903121MG
ABT-325 is a humanized antibody expressed in CHO that targets IL-18 ABT-325 has a huIgG1 type heavy chain and a hu type light chain with a predicted molecular weight (MW) of 146 28 kDa The isotype control for ABT-325 can refer to Human IgG1 kappa Isotype Control (HY-P99001)