Learn More
Medchemexpress LLC HY-13998 100mg Medchemexpress, ABT-333 Dasabuvir CAS:1132935-63-7 Purity:98%
SDP
Catalog No. 501638084
Encompass_Preferred
Medchemexpress,HY-13998 100mg ABT-333 Dasabuvir CAS:1132935-63-7 Formula:C26H27N3O5S ABT-333 is a nonnucleoside inhibitor of the RNA-dependent RNA polymerase encoded by the HCV NS5B gene, inhibits recombinant NS5B polymerases derived from HCV genotype 1a and 1b clinical isolates, with IC50 between 2.2 and 10.7 nM. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-8084 Supplier Medchemexpress LLC Supplier No. HY13998100MG
Medchemexpress,HY-13998 100mg ABT-333 Dasabuvir CAS:1132935-63-7 Formula:C26H27N3O5S ABT-333 is a nonnucleoside inhibitor of the RNA-dependent RNA polymerase encoded by the HCV NS5B gene, inhibits recombinant NS5B polymerases derived from HCV genotype 1a and 1b clinical isolates, with IC50 between 2.2 and 10.7 nM. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.