Learn More
Medchemexpress LLC Abt-418 hydrochloride | 147388-83-8 | 99.4% | 202.68 | C9H15ClN2O | 10 MG
SDP
Catalog No. 5000242856
Encompass_Preferred
ABT-418 hydrochloride is the hydrochloride salt of a selective neuronal nicotinic acetylcholine receptor (nAChR) agonist used for preclinical research. It is supplied as a light brown to brown solid, with characterized purity and solubility suitable for formulation and biological testing.
  • Selective neuronal nicotinic acetylcholine receptor (nAChR) agonist.
  • High purity (99.4%).
  • Molecular weight 202.68.
  • Water solubility approximately 11.9 mg/mL; requires ultrasonic.
  • PBS dissolution up to 100 mg/mL for in vivo formulation.
  • Solid, light brown to brown appearance; store sealed away from moisture at 4°C.
  • Available in small research pack sizes (5 mg-100 mg).
Catalog No. 50-002-42856 Supplier Medchemexpress LLC Supplier No. HY105170B10MG
ABT-418 hydrochloride is the hydrochloride salt of a selective neuronal nicotinic acetylcholine receptor (nAChR) agonist used for preclinical research. It is supplied as a light brown to brown solid, with characterized purity and solubility suitable for formulation and biological testing.
  • Selective neuronal nicotinic acetylcholine receptor (nAChR) agonist.
  • High purity (99.4%).
  • Molecular weight 202.68.
  • Water solubility approximately 11.9 mg/mL; requires ultrasonic.
  • PBS dissolution up to 100 mg/mL for in vivo formulation.
  • Solid, light brown to brown appearance; store sealed away from moisture at 4°C.
  • Available in small research pack sizes (5 mg-100 mg).