Learn More
Medchemexpress LLC Abt-418 hydrochloride | 147388-83-8 | MFCD11045960 | 99.4% | 202.68 | C9H15ClN2O | 25 MG
SDP
Catalog No. 5000242235
Encompass_Preferred
ABT-418 hydrochloride is the hydrochloride salt of a potent, selective nicotinic acetylcholine receptor (nAChR) agonist used in preclinical research on cholinergic signaling, cognition, and anxiolytic effects. Supplied as a high-purity research chemical for laboratory use only, it is suitable for in vitro and in vivo studies investigating nAChR pharmacology.
  • Potent, selective nAChR agonist.
  • Hydrochloride salt form for improved stability and handling.
  • High purity (99.4%).
  • Molecular weight 202.68 and chemical formula C9H15ClN2O.
  • Available in small research pack sizes, including 25 mg.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-42235 Supplier Medchemexpress LLC Supplier No. HY105170B25MG
ABT-418 hydrochloride is the hydrochloride salt of a potent, selective nicotinic acetylcholine receptor (nAChR) agonist used in preclinical research on cholinergic signaling, cognition, and anxiolytic effects. Supplied as a high-purity research chemical for laboratory use only, it is suitable for in vitro and in vivo studies investigating nAChR pharmacology.
  • Potent, selective nAChR agonist.
  • Hydrochloride salt form for improved stability and handling.
  • High purity (99.4%).
  • Molecular weight 202.68 and chemical formula C9H15ClN2O.
  • Available in small research pack sizes, including 25 mg.
  • For research use only; not for human or clinical use.