Learn More
Medchemexpress LLC Abt-418 (hydrochloride) | 147388-83-8 | MFCD11045960 | 99.4% | 202.68 g/mol | C9H15ClN2O | 5 MG
SDP
Catalog No. 5000242685
Encompass_Preferred
ABT-418 hydrochloride is a potent, selective agonist of neuronal nicotinic acetylcholine receptors (nAChRs) with reported cognitive-enhancing and anxiolytic activity. Supplied as a hydrochloride salt, it is intended for preclinical research applications, including studies related to Alzheimer's disease.
  • Potent, selective nAChR agonist.
  • Reported cognitive-enhancing and anxiolytic activity in preclinical models.
  • High purity (99.4%) for reliable experimental results.
  • Solid form, light brown to brown, convenient for handling.
  • Storage guidance provided for long-term stability.
  • Synonyms provided to aid literature and database searches.
Catalog No. 50-002-42685 Supplier Medchemexpress LLC Supplier No. HY105170B5MG
ABT-418 hydrochloride is a potent, selective agonist of neuronal nicotinic acetylcholine receptors (nAChRs) with reported cognitive-enhancing and anxiolytic activity. Supplied as a hydrochloride salt, it is intended for preclinical research applications, including studies related to Alzheimer's disease.
  • Potent, selective nAChR agonist.
  • Reported cognitive-enhancing and anxiolytic activity in preclinical models.
  • High purity (99.4%) for reliable experimental results.
  • Solid form, light brown to brown, convenient for handling.
  • Storage guidance provided for long-term stability.
  • Synonyms provided to aid literature and database searches.