Learn More
Medchemexpress LLC Abt-510 acetate | 442526-87-6 | 99.6% | 1054.28 g·mol⁻¹ | C48H87N13O13 | 5 MG
SDP
Catalog No. 5000222008
Encompass_Preferred
ABT-510 acetate is a synthetic thrombospondin-1 (TSP-1) mimetic peptide with anti-angiogenic activity used in preclinical research. It induces apoptosis and inhibits tumor growth in animal models, and is supplied at high purity for biochemical and in vivo studies.
  • Anti-angiogenic thrombospondin-1 mimetic peptide.
  • Induces apoptosis and inhibits tumor growth in animal models.
  • High purity (99.6%).
  • Suitable for in vitro and in vivo angiogenesis and apoptosis studies.
  • Available in small-scale quantities for preclinical experiments.
  • Includes supporting documents such as datasheet, SDS, COA, and LCMS.
Catalog No. 50-002-22008 Supplier Medchemexpress LLC Supplier No. HY13545B5MG
ABT-510 acetate is a synthetic thrombospondin-1 (TSP-1) mimetic peptide with anti-angiogenic activity used in preclinical research. It induces apoptosis and inhibits tumor growth in animal models, and is supplied at high purity for biochemical and in vivo studies.
  • Anti-angiogenic thrombospondin-1 mimetic peptide.
  • Induces apoptosis and inhibits tumor growth in animal models.
  • High purity (99.6%).
  • Suitable for in vitro and in vivo angiogenesis and apoptosis studies.
  • Available in small-scale quantities for preclinical experiments.
  • Includes supporting documents such as datasheet, SDS, COA, and LCMS.