Learn More
Medchemexpress LLC Abt-510 acetate | 442526-87-6 | 99.6% | C48H87N13O13 | 10MG
SDP
Catalog No. 5000259812
Encompass_Preferred
ABT-510 acetate is a synthetic thrombospondin-1 mimetic nonapeptide with anti-angiogenic and pro-apoptotic activity used in preclinical cancer and inflammatory disease research. It has demonstrated inhibition of tumor angiogenesis in ovarian cancer models and reduces inflammatory angiogenesis in animal studies. Supplied as a white to off-white solid powder with defined sequence and molecular characterization.
  • Anti-angiogenic and pro-apoptotic activity demonstrated in preclinical models.
  • Suitable for cancer and inflammatory disease research applications.
  • High purity as reported by supplier, suitable for analytical use.
  • White to off-white solid powder form for convenient storage and handling.
  • Characterized sequence and molecular weight support analytical verification.
Catalog No. 50-002-59812 Supplier Medchemexpress LLC Supplier No. HY13545B10MG
ABT-510 acetate is a synthetic thrombospondin-1 mimetic nonapeptide with anti-angiogenic and pro-apoptotic activity used in preclinical cancer and inflammatory disease research. It has demonstrated inhibition of tumor angiogenesis in ovarian cancer models and reduces inflammatory angiogenesis in animal studies. Supplied as a white to off-white solid powder with defined sequence and molecular characterization.
  • Anti-angiogenic and pro-apoptotic activity demonstrated in preclinical models.
  • Suitable for cancer and inflammatory disease research applications.
  • High purity as reported by supplier, suitable for analytical use.
  • White to off-white solid powder form for convenient storage and handling.
  • Characterized sequence and molecular weight support analytical verification.