Learn More
TARGETMOL CHEMICALS INC ABT-510 acetate 5MG
SDP
Catalog No. 502878229
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. ABT-510 acetate is an endogenous anti-angiogenic TSP peptide inhibitor, a thrombospondin analog, with anti-inflammatory, anti-cancer and anti-angiogenic activity that induces apoptosis and inhibits ovarian tumor growth in an orthotopic, syngeneic model of epithelial ovarian cancer.ABT-510 acetate reduces angiogenesis and inflammatory responses in a mouse model of inflammatory bowel disease. ABT-510 acetate reduces angiogenesis and inflammation in mouse models of inflammatory bowel disease (IBD) and can be used in cancer research, particularly in epithelial ovarian cancer, as well as in inflammatory bowel disease (IBD). Purity 95.07%
Catalog No. 50-287-8229 Supplier TARGETMOL CHEMICALS INC Supplier No. T249975MG
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. ABT-510 acetate is an endogenous anti-angiogenic TSP peptide inhibitor, a thrombospondin analog, with anti-inflammatory, anti-cancer and anti-angiogenic activity that induces apoptosis and inhibits ovarian tumor growth in an orthotopic, syngeneic model of epithelial ovarian cancer.ABT-510 acetate reduces angiogenesis and inflammatory responses in a mouse model of inflammatory bowel disease. ABT-510 acetate reduces angiogenesis and inflammation in mouse models of inflammatory bowel disease (IBD) and can be used in cancer research, particularly in epithelial ovarian cancer, as well as in inflammatory bowel disease (IBD). Purity 95.07%