Learn More
TARGETMOL CHEMICALS INC ABT-639 10MG
SDP
Catalog No. 502878825
Encompass_Preferred
Also available in 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. ABT-639 is a potent and selective T-type calcium channel blocker.
Catalog No. 50-287-8825 Supplier TARGETMOL CHEMICALS INC Supplier No. T370310MG
Also available in 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. ABT-639 is a potent and selective T-type calcium channel blocker.