Learn More
Apexbio Technology LLC ABT 702 dihydrochloride 1188890-28-9 10mg
SDP
Catalog No. 5000089669
Encompass_Preferred
ABT 702 dihydrochloride is a small-molecule inhibitor targeting adenosine kinase (AK) It is designed to inhibit AK the primary enzyme responsible for phosphorylation and metabolic inactivation of endogenous adenosine thereby elevating extracellular adenosine levels and modulating adenosine-mediated signaling ABT 702 dihydrochloride exerts its biological activity primarily through selective inhibition of AK In vitro studies indicate that ABT 702 potently inhibits AK activity and adenosine phosphorylation in intact cells Based on these pharmacological properties ABT 702 dihydrochloride holds research potential in inflammation and pain management particularly within experimental models of inflammatory and neuropathic pain
Catalog No. 50-000-89669 Supplier Apexbio Technology LLC Supplier No. B529810
ABT 702 dihydrochloride is a small-molecule inhibitor targeting adenosine kinase (AK) It is designed to inhibit AK the primary enzyme responsible for phosphorylation and metabolic inactivation of endogenous adenosine thereby elevating extracellular adenosine levels and modulating adenosine-mediated signaling ABT 702 dihydrochloride exerts its biological activity primarily through selective inhibition of AK In vitro studies indicate that ABT 702 potently inhibits AK activity and adenosine phosphorylation in intact cells Based on these pharmacological properties ABT 702 dihydrochloride holds research potential in inflammation and pain management particularly within experimental models of inflammatory and neuropathic pain