Learn More
Medchemexpress LLC Abt-702 dihydrochloride | 1188890-28-9 | 99.6% | 536.25 g/mol | C22H21BrCl2N6O | 50 MG
SDP
Catalog No. 5000247586
Encompass_Preferred
ABT-702 dihydrochloride is a potent, selective adenosine kinase (AK) inhibitor (IC50 = 1.7 nM) supplied as the dihydrochloride salt in solid form for laboratory research. The compound is reported with high purity and defined solubility and storage recommendations for in vitro and in vivo studies.
  • Potent adenosine kinase inhibitor with IC50 of 1.7 nM.
  • High purity reported at 99.64%.
  • Solid dihydrochloride salt suitable for biochemical assays.
  • Soluble in DMSO (≥ 33.33 mg/mL) and formulatable for in vivo dosing.
  • Recommended sealed storage at 4°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Available in multiple pack sizes including 50 MG.
Catalog No. 50-002-47586 Supplier Medchemexpress LLC Supplier No. HY10316150MG
ABT-702 dihydrochloride is a potent, selective adenosine kinase (AK) inhibitor (IC50 = 1.7 nM) supplied as the dihydrochloride salt in solid form for laboratory research. The compound is reported with high purity and defined solubility and storage recommendations for in vitro and in vivo studies.
  • Potent adenosine kinase inhibitor with IC50 of 1.7 nM.
  • High purity reported at 99.64%.
  • Solid dihydrochloride salt suitable for biochemical assays.
  • Soluble in DMSO (≥ 33.33 mg/mL) and formulatable for in vivo dosing.
  • Recommended sealed storage at 4°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Available in multiple pack sizes including 50 MG.