Learn More
Medchemexpress LLC Abt-702 dihydrochloride | 1188890-28-9 | 99.8% | 536.25 g/mol | C22H21BrCl2N6O | 100 MG
SDP
Catalog No. 5000246449
Encompass_Preferred
ABT-702 dihydrochloride is a potent, non-nucleoside adenosine kinase (AK) inhibitor (IC50 = 1.7 nM), supplied as the dihydrochloride salt for research use. It is used in biochemical and cellular studies to modulate adenosine kinase activity.
  • Potent adenosine kinase inhibitor with IC50 = 1.7 nM.
  • Dihydrochloride salt form for improved stability and solubility.
  • Molecular formula C22H21BrCl2N6O; molecular weight 536.25 g/mol.
  • Intended for research use in biochemical and cellular assays.
  • Store sealed away from moisture; in solvent, -80°C (up to 6 months) or -20°C (up to 1 month).
Catalog No. 50-002-46449 Supplier Medchemexpress LLC Supplier No. HY103161100MG
ABT-702 dihydrochloride is a potent, non-nucleoside adenosine kinase (AK) inhibitor (IC50 = 1.7 nM), supplied as the dihydrochloride salt for research use. It is used in biochemical and cellular studies to modulate adenosine kinase activity.
  • Potent adenosine kinase inhibitor with IC50 = 1.7 nM.
  • Dihydrochloride salt form for improved stability and solubility.
  • Molecular formula C22H21BrCl2N6O; molecular weight 536.25 g/mol.
  • Intended for research use in biochemical and cellular assays.
  • Store sealed away from moisture; in solvent, -80°C (up to 6 months) or -20°C (up to 1 month).