Learn More
Medchemexpress LLC Abt-702 dihydrochloride | 1188890-28-9 | MFCD03452809 | 99.6% | 536.25 g·mol-1 | C22H21BrCl2N6O | 10 MG
SDP
Catalog No. 5000247688
Encompass_Preferred
ABT-702 dihydrochloride is the dihydrochloride salt of ABT-702, a potent non-nucleoside adenosine kinase (AK) inhibitor (reported IC50 = 1.7 nM). It is provided as a solid research reagent for biochemical assays, pharmacological studies, and in vivo models involving adenosine metabolism and signaling.
  • Potent adenosine kinase inhibitor with reported IC50 = 1.7 nM.
  • Dihydrochloride salt form for improved handling and stability.
  • Molecular weight 536.25 g·mol-1; formula C22H21BrCl2N6O.
  • High purity (≈99.6%) suitable for research applications.
  • Supplied in small-scale quantities for laboratory use (typical 10 mg).
  • Commonly used in enzyme inhibition assays and in vivo pharmacology studies.
Catalog No. 50-002-47688 Supplier Medchemexpress LLC Supplier No. HY10316110MG
Add to Cart
Add to Cart
ABT-702 dihydrochloride is the dihydrochloride salt of ABT-702, a potent non-nucleoside adenosine kinase (AK) inhibitor (reported IC50 = 1.7 nM). It is provided as a solid research reagent for biochemical assays, pharmacological studies, and in vivo models involving adenosine metabolism and signaling.
  • Potent adenosine kinase inhibitor with reported IC50 = 1.7 nM.
  • Dihydrochloride salt form for improved handling and stability.
  • Molecular weight 536.25 g·mol-1; formula C22H21BrCl2N6O.
  • High purity (≈99.6%) suitable for research applications.
  • Supplied in small-scale quantities for laboratory use (typical 10 mg).
  • Commonly used in enzyme inhibition assays and in vivo pharmacology studies.