Learn More
Medchemexpress LLC Abt-724 trihydrochloride | 587870-77-7 | 99.1% | 402.75 g/mol | C17H22Cl3N5 | 50 MG
SDP
Catalog No. 5000245197
Encompass_Preferred
ABT-724 trihydrochloride is a potent, selective dopamine D4 receptor agonist supplied as a white to off-white solid for research use. It shows low-nanomolar activity at human D4 receptors (EC50 12.4 nM) and has been investigated for potential treatment of erectile dysfunction. Store sealed and away from moisture; in solution, store at -80°C (up to 6 months) or -20°C (up to 1 month).
  • High purity suitable for research applications.
  • Potent dopamine D4 receptor agonist with low-nanomolar EC50.
  • White to off-white solid, easy to handle.
  • Available in multiple package sizes to suit experiments.
  • Stable when stored sealed; follow recommended temperature guidelines.
Catalog No. 50-002-45197 Supplier Medchemexpress LLC Supplier No. HY10340950MG
ABT-724 trihydrochloride is a potent, selective dopamine D4 receptor agonist supplied as a white to off-white solid for research use. It shows low-nanomolar activity at human D4 receptors (EC50 12.4 nM) and has been investigated for potential treatment of erectile dysfunction. Store sealed and away from moisture; in solution, store at -80°C (up to 6 months) or -20°C (up to 1 month).
  • High purity suitable for research applications.
  • Potent dopamine D4 receptor agonist with low-nanomolar EC50.
  • White to off-white solid, easy to handle.
  • Available in multiple package sizes to suit experiments.
  • Stable when stored sealed; follow recommended temperature guidelines.