Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Abt-724 trihydrochloride | 587870-77-7 | MFCD08703101 | 99.1% | 402.75 g/mol | C17H22Cl3N5 | 25 MG
SDP

Catalog No. 5000244712
Encompass_Preferred

ABT-724 trihydrochloride is a research-grade small molecule dopamine D4 receptor agonist supplied as the trihydrochloride salt. It is intended for pharmacology and neuroscience research and is provided as a high-purity solid.

  • Potent and selective dopamine D4 receptor agonist (EC50 12.4 nM).
  • High purity (~99.1% by HPLC).
  • Molecular weight 402.75 g/mol; formula C17H22Cl3N5.
  • Supplied as a 25 mg research reagent.
  • Appearance: solid; store sealed and away from moisture.
  • In solvent stability: -80°C (up to 6 months), -20°C (up to 1 month).

Catalog No. 50-002-44712 Supplier Medchemexpress LLC Supplier No. HY10340925MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ABT-724 trihydrochloride is a research-grade small molecule dopamine D4 receptor agonist supplied as the trihydrochloride salt. It is intended for pharmacology and neuroscience research and is provided as a high-purity solid.

  • Potent and selective dopamine D4 receptor agonist (EC50 12.4 nM).
  • High purity (~99.1% by HPLC).
  • Molecular weight 402.75 g/mol; formula C17H22Cl3N5.
  • Supplied as a 25 mg research reagent.
  • Appearance: solid; store sealed and away from moisture.
  • In solvent stability: -80°C (up to 6 months), -20°C (up to 1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.