Learn More
Apexbio Technology LLC ABT-737 852808-04-9 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062207
Encompass_Preferred
ABT-737 (CAS 852808-04-9) is a small molecule inhibitor targeting the BCL-2 protein family exhibiting high binding affinity toward the anti-apoptotic proteins BCL-2 BCL-X L and BCL-w ABT-737 disrupts BCL-2/BAX protein interactions leading to apoptosis primarily via the BAK-mediated intrinsic pathway independent of BIM In preclinical studies ABT-737 demonstrated single-agent antitumor activity in lymphoma multiple myeloma and small-cell lung cancer models Additionally research indicates activity against acute myeloid leukemia (AML) cells progenitor cells and leukemia stem cells while sparing normal hematopoietic populations highlighting its potential for therapeutic research in hematologic malignancies
Catalog No. 50-000-62207 Supplier Apexbio Technology LLC Supplier No. A81935.1
 Restricted Item
Add to Cart
Add to Cart
ABT-737 (CAS 852808-04-9) is a small molecule inhibitor targeting the BCL-2 protein family exhibiting high binding affinity toward the anti-apoptotic proteins BCL-2 BCL-X L and BCL-w ABT-737 disrupts BCL-2/BAX protein interactions leading to apoptosis primarily via the BAK-mediated intrinsic pathway independent of BIM In preclinical studies ABT-737 demonstrated single-agent antitumor activity in lymphoma multiple myeloma and small-cell lung cancer models Additionally research indicates activity against acute myeloid leukemia (AML) cells progenitor cells and leukemia stem cells while sparing normal hematopoietic populations highlighting its potential for therapeutic research in hematologic malignancies