Learn More
Medchemexpress LLC HY-50907 200mg , ABT-737 CAS:852808-04-9 Purity:98%
SDP
Catalog No. 501492557
Encompass_Preferred
Medchemexpress, HY-50907 200mg ABT-737 CAS:852808-04-9 Formula:C42H45ClN6O5S2 EC50: 78.7 nM (Bcl-xL), 30.3 nM (Bcl-2), 197.8 nM (Bcl-w) Purity:98% ABT-737 is a BH3 mimetic inhibitor of  Bcl-xL ,  Bcl-2  and  Bcl-w  with  EC 50  of 78.7 nM, 30.3 nM and 197.8 nM in cell-free assays, respectively, and shows no inhibition against Mcl-1, Bcl-B or Bfl-1. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2557 Supplier Medchemexpress LLC Supplier No. HY50907200MG
Medchemexpress, HY-50907 200mg ABT-737 CAS:852808-04-9 Formula:C42H45ClN6O5S2 EC50: 78.7 nM (Bcl-xL), 30.3 nM (Bcl-2), 197.8 nM (Bcl-w) Purity:98% ABT-737 is a BH3 mimetic inhibitor of  Bcl-xL ,  Bcl-2  and  Bcl-w  with  EC 50  of 78.7 nM, 30.3 nM and 197.8 nM in cell-free assays, respectively, and shows no inhibition against Mcl-1, Bcl-B or Bfl-1. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.