Learn More
Apexbio Technology LLC ABT-737, 50mg. Cas: 852808-04-9 MFCD: MFCD12756212. Bcl-2 inhibitor
SDP
Catalog No. 501905072
Encompass_Preferred
ABT-737 is a novel and potent inhibitor of B-cell lymphoma 2 (BCL-2) family proteins, which are critical for cell survival and overexpressed in many tumor cells, with high affinity towards BCL-XL, BCL-2, and BCL-w but no affinity towards less homologous proteins, such as BCL-B, MCL-1, and A1. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5072 Supplier Apexbio Technology LLC Supplier No. A819350
ABT-737 is a novel and potent inhibitor of B-cell lymphoma 2 (BCL-2) family proteins, which are critical for cell survival and overexpressed in many tumor cells, with high affinity towards BCL-XL, BCL-2, and BCL-w but no affinity towards less homologous proteins, such as BCL-B, MCL-1, and A1. Other sizes are available. Please inqury us for quote.