Learn More
Selleck Chemical LLC ABT-751 (E7010) 10mg 141430-65-1
SDP
Catalog No. 507807
Encompass_Preferred
ABT-751 (E7010) binds to the colchicine site on ?-tubulin and inhibits polymerization of microtubules, not a substrate for the MDR transporter and is active against cell lines resistant to vincristine, doxorubicin, and cisplatin. Phase 1/2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-780-7 Supplier Selleck Chemical LLC Supplier No. S116510MG
ABT-751 (E7010) binds to the colchicine site on ?-tubulin and inhibits polymerization of microtubules, not a substrate for the MDR transporter and is active against cell lines resistant to vincristine, doxorubicin, and cisplatin. Phase 1/2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.