Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABT 751, Tocris Bioscience™

Catalog No. 413810
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-381-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 41-381-0 Supplier R&D Systems Supplier No. 4138/10
This item is not returnable. View return policy

Inhibitor of microtubule polymerization; antimitotic and antitumor

Antimitotic agent, inhibits microtubule polymerization. Binds to β-tubulin on the colchine site; blocks cell cycle at G2M phase and induces apoptosis. Exhibits antitumor activity. Also displays activity in cell lines resistant to vincristine, doxorubicin and cisplatin. Orally bioavailable.

Chemical Identifiers

CAS 141430-65-1
Molecular Formula C18H17N3O4S
Molecular Weight (g/mol) 371.411
InChI Key URCVCIZFVQDVPM-UHFFFAOYSA-N
Synonym unii-wdt5v5ob9f, wdt5v5ob9f, chembl20684, n-2-4-hydroxyphenyl amino-3-pyridyl-4-methoxybenzenesulfonamide, n-2-4-hydroxyphenyl amino-3-pyridinyl-4-methoxybenzenesulfonamide, n-2-4-hydroxyphenyl amino pyridin-3-yl-4-methoxybenzenesulfonamide, benzenesulfonamide,n-2-4-hydroxyphenyl amino-3-pyridinyl-4-methoxy, n-2-4-hydroxyphenylamino pyridin-3-yl-4-methoxybenzenesulfonamide, benzenesulfonamide, n-2-4-hydroxyphenyl amino-3-pyridinyl-4-methoxy
PubChem CID 3035714
IUPAC Name N-[2-(4-hydroxyanilino)pyridin-3-yl]-4-methoxybenzenesulfonamide
SMILES COC1=CC=C(C=C1)S(=O)(=O)NC2=C(N=CC=C2)NC3=CC=C(C=C3)O

Specifications

Quantity 10 mg
Formula Weight 371.41
Percent Purity >98%
Chemical Name or Material ABT 751
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.