Learn More
Apexbio Technology LLC ABT-888 (Veliparib), 50mg. Cas: 912444-00-9 MFCD: MFCD16661059. Potent PARP inhibitor
SDP
Supplier:  Apexbio Technology LLC A300250
Encompass_Preferred
ABT-888, also named as Veliparib, is poly (ADP-ribose) polymerase (PARP) inhibitor and has demonstrated excellent in vivo efficacy in a broad spectrum of preclinical tumor models in combination with a variety of cytotoxic agents. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4772