Learn More
Aviva Systems Biology Corp Abt1 Antibody - C-terminal region (ARP37211_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37211_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Abt1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the c terminal region of mouse Abt1. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Mouse, Pig, Rabbit, Rat
Catalog No. 50-284-595