Learn More
Aviva Systems Biology Corp Abt1 Antibody - middle region (ARP37210_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37210_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Abt1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of mouse Abt1. Alternate names: -ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-594