Learn More
Aviva Systems Biology Corp ABT1 Antibody - N-terminal region (ARP31915_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31915_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ABT1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ABT1. Alternate names: hABT1ApplicationWB. Species reactivity: Cow, Dog, Human, Rabbit
Catalog No. 50-014-54369