Learn More
TARGETMOL CHEMICALS INC ABT530 10MG
SDP
Catalog No. 502579807
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. ABT530 (Pibrentasvir) is an HCV NS5A inhibitor with EC50s ranging from 1.4-5.0 pM against HCV replicons containing NS5A from genotypes 1-6. purity: 99%
Catalog No. 50-257-9807 Supplier TARGETMOL CHEMICALS INC Supplier No. T512710MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. ABT530 (Pibrentasvir) is an HCV NS5A inhibitor with EC50s ranging from 1.4-5.0 pM against HCV replicons containing NS5A from genotypes 1-6. purity: 99%