Learn More
Seracare Life Sciences Inc ABTS 2-C ELISA HRP SUBSTRATE
Supplier:  Seracare Life Sciences Inc 51200032
Encompass_Preferred
A good, all-purpose substrate that offers the widest working range. Ideal when low background is required and moderate sensitivity is acceptable. ABTS develops a blue-green color in the presence of peroxidase labeled conjugates measurable between 405 and 410 nm. No change in color when reaction is stopped. Recommended for ELISA applications. Two-component format that contains 2, 2'-azino-di(3-ethylbenzthiazoline-6-sulfonate), simply mix and use. Has longer shelf life than single component format.
Catalog No. 50-674-08