Learn More
Seracare Life Sciences Inc ABTS 2-C ELISA HRP SUBSTRATE
Supplier:  Seracare Life Sciences Inc 51200033
Encompass_Preferred
KPL 3 x 450 mL ABTS Solution A, 3 x 450 mL ABTS Solution B. Product Highlights:Widest linear range of peroxidase colorimetric ELISA substrates;Convenient, mix and use solutions;Promotes lower background levels. A good, all-purpose substrate that offers the widest working range. Ideal when low background is required and moderate sensitivity is acceptable. ABTS develops a blue-green color in the presence of peroxidase labeled conjugates measurable between 405 and 410 nm. No change in color when reaction is stopped. Recommended for ELISA applications. Two-component format that contains 2, 2'-azino-di(3-ethylbenzthiazoline-6-sulfonate), simply mix and use. Has longer shelf life than single component format.
Catalog No. 50-674-09