Learn More
Selleck Chemical LLC ABTS E2917-100MG
SDP
Catalog No. 5000040302
Encompass_Preferred
ABTS (ABTS diammonium salt AzBTS-(NH4)2) is a substrate for horseradish peroxidase (HRP) conjugate It has commonly been used to assess antioxidant capacity in the Trolox equivalent antioxidant capacity (TEAC) assay
Catalog No. 50-000-40302 Supplier Selleck Chemical LLC Supplier No. E2917100MG
ABTS (ABTS diammonium salt AzBTS-(NH4)2) is a substrate for horseradish peroxidase (HRP) conjugate It has commonly been used to assess antioxidant capacity in the Trolox equivalent antioxidant capacity (TEAC) assay