Learn More
Medchemexpress LLC HY-15902 500mg Medchemexpress, ABTS (diammonium salt) CAS:30931-67-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15902500MG
Encompass_Preferred
Medchemexpress, HY-15902 500mg ABTS (diammonium salt) CAS:30931-67-0 ABTS diammonium salt is a substrate for horseradish peroxidase (HRP) conjugate. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1822