Learn More
Abcam ABTS diammonium salt, chromogenic pero x idase substrate, 5G
Catalog No. 502704696
Encompass_Preferred
MW 548.7 g/mol, Purity >=98%. Chromogenic peroxidase substrate widely utilized in enzyme-linked immunosorbent assay (ELISA) procedures. Produces a soluble end product that is green in color and can be read spectrophotometrically at 405 nm. Also used in the food industry to measure the antioxidant capacities of foods.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4696 Supplier Abcam Supplier No. AB1463435G
MW 548.7 g/mol, Purity >=98%. Chromogenic peroxidase substrate widely utilized in enzyme-linked immunosorbent assay (ELISA) procedures. Produces a soluble end product that is green in color and can be read spectrophotometrically at 405 nm. Also used in the food industry to measure the antioxidant capacities of foods.
The product is subject to the following: Abcam Restricted Use Statement