Learn More
Rockland Immunochemicals ABTS ELISA PEROXIDASE SUBSTRATE - ABTS-100
SDP
Supplier:  Rockland Immunochemicals ABTS100
Encompass
ABTS ELISA Peroxidase Substrate (2,2'-azino-bis(3-ethylbenzothiazoline-6-sulphonic acid)) is a chromogenic substrate use to visualize reactivity of certain enzymes, particularly in immunosorbent assays. ABTS ELISA Peroxidase Substrate is commonly used with peroxidases, but is amenable to many other enzymes. In the presence of active enzyme, the substrate becomes a soluble green product detectable at 405nm.
Catalog No. 50-105-8862