Learn More
Seracare Life Sciences Inc ABTS HRP STOP SOLUTION
Catalog No. 5067442
Encompass_Preferred
2 x 100 mL ABTS Peroxidase Stop Solution Product Highlights - Allows accurate end-point determinations when used with ABTS substrates - Plates may be read up to 30 minutes after stopping. - Substrate color does not change after addition of ABTS Stop Solution ABTS Peroxidase Stop Solution contains 5% sodium dodecyl sulfate (SDS) in water. This solution is specifically designed for use with KPL’s ABTS Peroxidase Substrates (both 1- and 2-component) to stop development in ELISA plate wells.
This item is not returnable. View return policy
Catalog No. 50-674-42 Supplier Seracare Life Sciences Inc Supplier No. 51500017
2 x 100 mL ABTS Peroxidase Stop Solution Product Highlights - Allows accurate end-point determinations when used with ABTS substrates - Plates may be read up to 30 minutes after stopping. - Substrate color does not change after addition of ABTS Stop Solution ABTS Peroxidase Stop Solution contains 5% sodium dodecyl sulfate (SDS) in water. This solution is specifically designed for use with KPL’s ABTS Peroxidase Substrates (both 1- and 2-component) to stop development in ELISA plate wells.