Learn More
Bioworld ABTS, Diammonium Salt, 5 g
Catalog No. 50198844
Encompass_Preferred
ABTS, Diammonium Salt Also known as 2,2'-Azino-bis(3-ethylbenzthiazoline-6-sulfonic acid). Technical Information:Ultra Pure GradePurity: >98.0%Em (225nm, H2O): >48.150Solubility (H2O): 5%Store: Frozen
Catalog No. 50-198-844 Supplier Bioworld Supplier No. 215300423
ABTS, Diammonium Salt Also known as 2,2'-Azino-bis(3-ethylbenzthiazoline-6-sulfonic acid). Technical Information:Ultra Pure GradePurity: >98.0%Em (225nm, H2O): >48.150Solubility (H2O): 5%Store: Frozen