Learn More
Adam Equipment ABW 8 - Aqua Washdown Scales - Capacity: 18lb / 8kg, Readability: 0.002lb / 1g
Supplier:  Adam Equipment ABW 8
Encompass_Preferred
Aqua Washdown Scales from Adam Equipment deliver durability and precision for food processing and industrial environments. With an IP67 rating, they are protected from dust and water immersion, making them ideal for frequent washdowns. The stainless steel pan ensures easy cleaning and long-lasting performance.Dual LED displays allow multiple operators to work simultaneously, while color-changing indicators simplify checkweighing. The scales also support percentage weighing and parts counting for versatile use. Fast one-second stabilization ensures quick, accurate results.Powered by AC or a rechargeable battery with up to 55 hours of operation, Aqua scales are built for flexibility. Features like auto power-off, overload protection, a sealed keypad, and heavy-duty rubber feet enhance reliability and longevity. With clear visibility and zero-tracking accuracy, Aqua scales provide a dependable, efficient weighing solution.
Catalog No. NC3964943