Learn More
Medchemexpress LLC HY-117632 10mg Medchemexpress, ABX-1431 CAS:1446817-84-0 Purity:>98%
SDP
Catalog No. 501932525
Encompass_Preferred
Medchemexpress, HY-117632 10mg ABX-1431 CAS:1446817-84-0 ABX-1431 is a highly potent, selective, and orally available, CNS-penetrant monoacylglycerol lipase ( MAGL ) inhibitor with an IC 50 of 14 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2525 Supplier Medchemexpress LLC Supplier No. HY11763210MG
Medchemexpress, HY-117632 10mg ABX-1431 CAS:1446817-84-0 ABX-1431 is a highly potent, selective, and orally available, CNS-penetrant monoacylglycerol lipase ( MAGL ) inhibitor with an IC 50 of 14 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.