Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HORIBA Hematology Reagent ABX Minidil LMG, Blood Cell Counting
GSA_VA

Catalog No. 23450008 Shop All ABX Diagnostics, Inc. Products
Change view
Click to view available options
Quantity:
10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-450-008 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-450-008 Supplier HORIBA Supplier No. 1210802010
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with Micros 60 and ES60

Buffered isotonic solution for the determination of blood cell counting and the measurement of hematocrit on ABX blood cell counters

  1. Bottle
  2. Cubitainer
  3. For 18 parameter analyzer.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

For Use With (Equipment) ABX Micros 60 and ES60 Analyzer
Type ABX Minidil LMG
For Use With (Application) Blood Cell Counting
Quantity 10 L
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.