Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HORIBA ABX Minipack LMG

Catalog No. 23450005 Shop All ABX Diagnostics, Inc. Products
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-450-005 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-450-005 Supplier HORIBA Supplier No. 1210602050
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with Micros 60 and ES60

  1. Bottle
  2. Cubitainer
  3. For 18 parameter analyzer.

Specifications

For Use With (Equipment) Micros 60 and ES60
Type ABX Minipack LMG,
No. of Reactions 150
Quantity Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.