Learn More
HORIBA™ ABX Minocal Whole Blood Hematology Calibrator
Catalog No. 22899101 Shop All ABX Diagnostics, Inc. Products
Change view
Click to view available options
Quantity:
1 x 2.5 mL
Catalog No. Quantity
22-899-101 1 x 2.5 mL
1 options
This item is not returnable. View return policy
Catalog No. 22-899-101 Supplier HORIBA™ Supplier No. 5300100300
Add to Cart
Add to Cart
This item is not returnable. View return policy
Minocal Calibrator for whole blood hematology.
Intended Use
ABX Minocal is a multiparameter blood callibrator designed for use in the calibrations of HORIBA ABX SAS hematology blood cell counters using HORIBA ABX SAS reagents.
Summary
The WBC, RBC, HGB, HCT, and PLT parameters on instruments required calibration on a periodic basis. ABX Minocal is a stable preparation which can be used to calibrate the instrument. Calibrator values for ABX minocal have been obtained from replicate analyses on instruments which have been whole blood calibrated to values obtained from reference mathods.
Reagents
This whole blood reagent may contain any or all of the following: stabilized human or animal red blood cells, human, animal or simulated white blood cells and a platelet component in a preservative medium.
Specifications
Type Hematology Calibrator
Level 1
Quantity 1 x 2.5 mL
Form Liquid