Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HORIBA™ ABX Minotrol 16 Whole Blood Hematology Control (L, N, H)

Catalog No. 22899103 Shop All ABX Diagnostics, Inc. Products
Change view
Click to view available options
Quantity:
6 x 2.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-899-103 6 x 2.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-899-103 Supplier HORIBA™ Supplier No. 5300100306
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Minocal Control for whole blood hematology.

Intended Use

ABX Minocal 16 is a tri-level control intended for in vitro diagnostic use in monitoring the accuracy and precision on HORIBA ABX SAS hematology blood cell counters using HORIBA ABX SAS reagents.

Summary

It is an established laboratory procedure to use stable controls to monitor analytical mthods. ABX Minotrol 16 is a stable materia that provides a means of determining the accuracy and precision of hematology blood cell counters. It is handled in the same manner as a patient specimen. The assay tables are determined on validated instruments using the appropriate reagents.

Reagents

This whole blood reagent may contain any or all of the following: stabilized human red blood cells, a white blood cell component consisting of human and/or non-human analogs and a platelet component consisting of a human or non-human analog in a preservative medium.

Specifications

Sample Type Whole Blood
Type Hematology Control
Level 3
Quantity 6 x 2.5 mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.