Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HORIBA ABX Pentra RF Cal

Catalog No. 23601585 Shop All ABX Diagnostics, Inc. Products
Change view
Click to view available options
Quantity:
5 Pk. 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-601-585 5 Pk. 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-601-585 Supplier HORIBA Supplier No. 1220001618
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ABX Rheumatoid Factor Calibrator used to calibrate RF assay

  • Used to calibrate RF assay

Specifications

Certifications/Compliance CE Marked
Content And Storage 2°C to 10°C
Concentration 5 Concentrations: 10, 20, 40, 80, 120 IU/mL
Detectable Analytes Rheumatoid Factor
Sample Type Serum
Technology Immunoturbidimetric Assay
Type Calibrator
Level 5 Levels: 10, 20, 40, 80, 120
Quantity 5 Pk. 1 mL
Source Human
Stability 5 weeks (at 2°C to 10°C)
Sterility Non-sterile
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.