Learn More
Medchemexpress LLC HY-100870 5mg Medchemexpress, ABX464 CAS:1258453-75-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1008705MG
Encompass_Preferred
Medchemexpress, HY-100870 5mg ABX464 CAS:1258453-75-6 ABX464 is a potent anti-HIV agent. ABX464 inhibits HIV-1 replication in stimulated peripheral blood mononuclear cells (PBMCs) with an IC 50 ranging between 0.1 μM and 0.5 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1977