Learn More
Carbosynth LLC. Abz-Ala-Pro-Glu-Glu-Ile-Met-Arg-Arg-Gln-EDDnp, 1mg, MW: 1456.59, Molecular formula: C61H93N21O19S
SDP
Supplier:  Carbosynth LLC. SNE3230V1MG
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Synonyms: 2-Aminobenzoyl-L-alanyl-L-prolyl-L-glutamyl-L-glutamyl-L-isoleucyl-L-methionyl-L-arginyl-L-arginyl-lglutamine2-(2,4-dinitrophenyl)aminoethylamide, Abz-APEEIMRRQ-EDDnp, Application: Fluorescence-Quenching Substrate for Human Neutrophil Elastase, Salt Form:, Trifluoroacetate Salt, Storage Temperature: -20C, References: B. Korkmaz, S. Attucci, T. Moreau, E. Godat, L. Juliano, and F. Gauthier, Am. J. Respir. Cell Mol. Biol., 30, 801 (2004)., B. Korkmaz, S. Attucci, M.A. Juliano, T. Kalupov, M.-L. Jourdan, L. Juliano, and F. Gauthier, Nat. Protoc., 3, 991 (2008).
Catalog No. 50-168-7138