Learn More
Carbosynth LLC. Abz-Glu-Pro-Phe-Trp-Glu-Asp-Gln-EDDnp (1 mg vial), 1ea, MW: 1277.3, Molecular formula: C59H68N14O19
SDP
Supplier:  Carbosynth LLC. SFR3231V1MG
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Synonyms: [Abz-EPFWEDQ-EDDnp]2-Aminobenzoyl-L-glutamyl-L-prolyl-L-phenylalanyl-L-tryptophyl-L-glutamyl-L-aspartyl-L-glutamine 2-(2,4-dinitrophenyl)aminoethyl-amide, Purity: , 98.0% (HPLC), Application: Fluorescence-Quenching Substrate for Human Neutrophil Cathepsin G, Salt Form:, Ammonium, Storage Temperature: -20 C, References: B. Korkmaz, S. Attucci, M.A. Juliano, T. Kalupov, M.-L. Jourdan, L. Juliano, and F. Gauthier, Nat. Protoc., 3, 991 (2008)., S. Attucci, B. Korkmaz, L. Juliano, E. Hazouard, C. Girardin, M. Brillard-Bourdet, S. Réhault, P. Anthonioz, and F. Gauthie, Biochem. J., 366, 965 (2002).
Catalog No. 50-168-7098