Learn More
Medchemexpress LLC Abz-FR-K(Dnp)-P-OH | 500799-61-1 | 99.48% | 831.87 | 10 MG
SDP
Catalog No. 5000423956
Encompass_Preferred
Abz-FR-K(Dnp)-P-OH is an angiotensin I-converting enzyme (ACE) substrate and an internally quenched fluorogenic substrate for real-time fluorescent assay.
  • Angiotensin I-converting enzyme (ACE) substrate
  • Internally quenched fluorogenic substrate
  • Used for real-time fluorescent assay
  • For research use only
Catalog No. 50-004-23956 Supplier Medchemexpress LLC Supplier No. HYP185310MG
Abz-FR-K(Dnp)-P-OH is an angiotensin I-converting enzyme (ACE) substrate and an internally quenched fluorogenic substrate for real-time fluorescent assay.
  • Angiotensin I-converting enzyme (ACE) substrate
  • Internally quenched fluorogenic substrate
  • Used for real-time fluorescent assay
  • For research use only