Learn More
CPC Scientific H-Abz-Gly-Ala-Lys(Ac)-Ala-Ala-Dap(DNP)-NH2 (trifluoroacetate salt) 1MG
SDP
Catalog No. 502109430
Encompass
This peptide is the FRET substrate for assaying histone deacetylase (HDAC) activity. The assay is based on the principle that the peptide is more sensitive to trypsin cleavage after deacetylation.SEQUENCE: H-Abz-Gly-Ala-Lys(Ac)-Ala-Ala-Dap(DNP)-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: Abz-GAK(Ac)-AA-Dap(Dnp)-NH2MOLECULAR FORMULA: C35H48N12O12MOLECULAR WEIGHT: 828.84STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [761443-02-1]SYNONYMS: RESEARCH AREA: MiscellaneousREFERENCES:B.E.Schultz et al., Biochemistry, 43, 11083 (2004)SKU(s): HIST-005A, HIST-005B
Catalog No. 50-210-9430 Supplier CPC Scientific Supplier No. HIST005A
This peptide is the FRET substrate for assaying histone deacetylase (HDAC) activity. The assay is based on the principle that the peptide is more sensitive to trypsin cleavage after deacetylation.SEQUENCE: H-Abz-Gly-Ala-Lys(Ac)-Ala-Ala-Dap(DNP)-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: Abz-GAK(Ac)-AA-Dap(Dnp)-NH2MOLECULAR FORMULA: C35H48N12O12MOLECULAR WEIGHT: 828.84STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [761443-02-1]SYNONYMS: RESEARCH AREA: MiscellaneousREFERENCES:B.E.Schultz et al., Biochemistry, 43, 11083 (2004)SKU(s): HIST-005A, HIST-005B