Learn More
Medchemexpress LLC Abz-Gly-Leu-Lys-Arg-Gly-Gly-3-(NO2)Tyr acetate | 99.9% | 913.98 (free base) | 5 MG
SDP
Catalog No. 5000398915
Encompass_Preferred
Abz-Gly-Leu-Lys-Arg-Gly-Gly-3-(NO2)Tyr acetate is the acetate salt form of Abz-Gly-Leu-Lys-Arg-Gly-Gly-3-(NO2)Tyr. It serves as a substrate for WNV NS2B NS3 protease and is used in the development of WNV protease substrate peptide selective inhibitors.
  • Substrate for WNV NS2B NS3 protease.
  • Utilized for the development of WNV protease substrate peptide selective inhibitor.
  • For research use only.
Catalog No. 50-003-98915 Supplier Medchemexpress LLC Supplier No. HYP5312A5MG
Abz-Gly-Leu-Lys-Arg-Gly-Gly-3-(NO2)Tyr acetate is the acetate salt form of Abz-Gly-Leu-Lys-Arg-Gly-Gly-3-(NO2)Tyr. It serves as a substrate for WNV NS2B NS3 protease and is used in the development of WNV protease substrate peptide selective inhibitors.
  • Substrate for WNV NS2B NS3 protease.
  • Utilized for the development of WNV protease substrate peptide selective inhibitor.
  • For research use only.